Learn More
Ace Glass, Inc. Flask, round bottom, two neck, 3L, 45/50 center, 24/40 side
SDP
Catalog No. 50978933
Encompass_Preferred
3L two neck, round bottom flask with standard joints. Center neck has a 45/50 joint, and the angled side neck has a 24/40 joint.
Catalog No. 50-978-933 Supplier Ace Glass, Inc. Supplier No. 6927160
3L two neck, round bottom flask with standard joints. Center neck has a 45/50 joint, and the angled side neck has a 24/40 joint.