Learn More
Ace Glass, Inc. Flask, round bottom, two neck, 500mL, 24/40 center, 10/30 side
SDP
Catalog No. 50973573
Encompass_Preferred
500mL two neck, round bottom flask with standard joints. Center neck has a 24/40 joint, and the angled side neck has a 10/30 joint.
Catalog No. 50-973-573 Supplier Ace Glass, Inc. Supplier No. 692720
500mL two neck, round bottom flask with standard joints. Center neck has a 24/40 joint, and the angled side neck has a 10/30 joint.