Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 500mL, 24/40 center, (2) 24/40, (1) 10/30 sides
SDP
Catalog No. 50973853
Encompass_Preferred
500ml round bottom flask with four necks, arranged three in-line, and one 10/30 joint at 90. Center neck has a 24/40 joint. Angled side necks consist of 24/40 joints.
Catalog No. 50-973-853 Supplier Ace Glass, Inc. Supplier No. 695304
500ml round bottom flask with four necks, arranged three in-line, and one 10/30 joint at 90. Center neck has a 24/40 joint. Angled side necks consist of 24/40 joints.