Learn More
Ace Glass, Inc. Flask, round bottom, two neck, 500mL, 24/40 center, 24/40 side
SDP
Catalog No. 50973574
Encompass_Preferred
500mL two neck, round bottom flask with standard joints. Center neck and angled side neck both have 24/40 joints.
Catalog No. 50-973-574 Supplier Ace Glass, Inc. Supplier No. 692722
500mL two neck, round bottom flask with standard joints. Center neck and angled side neck both have 24/40 joints.