Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 500mL, 29/42 center, 29/42 sides
SDP
Catalog No. 50973797
Encompass_Preferred
500mL round bottom flask with four necks, arranged three in-line and one at 90. Center neck and angled side necks have 29/42 joints.
Catalog No. 50-973-797 Supplier Ace Glass, Inc. Supplier No. 695203
500mL round bottom flask with four necks, arranged three in-line and one at 90. Center neck and angled side necks have 29/42 joints.