Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 500mL, 34/45 center, 24/40 sides
SDP
Catalog No. 50973798
Encompass_Preferred
500mL round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 34/45 joint, and angled side necks have 24/40 joints.
Catalog No. 50-973-798 Supplier Ace Glass, Inc. Supplier No. 695204
500mL round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 34/45 joint, and angled side necks have 24/40 joints.