Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 50L, 45/50 center, 45/50 sides
SDP
Catalog No. 50973816
Encompass_Preferred
50L round bottom flask with four necks, arranged three in-line and one at 90. Center neck and angled side necks have 45/50 joints.
Catalog No. 50-973-816 Supplier Ace Glass, Inc. Supplier No. 695279
50L round bottom flask with four necks, arranged three in-line and one at 90. Center neck and angled side necks have 45/50 joints.