Learn More
Ace Glass, Inc. Flask, round, three neck, 50mL, 14/10 cap-seal center and sides
SDP
Catalog No. 50976860
Encompass_Preferred
50mL round bottom flask with three necks. Center and angled side necks are 14/10 and feature externally threaded cap-seal joints supplied with 14.7-15mm holed caps, PTFE faced septa, and FETFE o-rings.
Catalog No. 50-976-860 Supplier Ace Glass, Inc. Supplier No. 946536
50mL round bottom flask with three necks. Center and angled side necks are 14/10 and feature externally threaded cap-seal joints supplied with 14.7-15mm holed caps, PTFE faced septa, and FETFE o-rings.