Learn More
Ace Glass, Inc. Flask, round, heavy duty, 50mL, 14/20 center, 14/20 side
SDP
Catalog No. 50976853
Encompass_Preferred
50mL heavy wall round bottom flask with (1) 14/20 standard taper center neck and (1) 14/20 standard taper angled side neck. Heavy wall provides mechanical shock resistance.
Catalog No. 50-976-853 Supplier Ace Glass, Inc. Supplier No. 9464208
50mL heavy wall round bottom flask with (1) 14/20 standard taper center neck and (1) 14/20 standard taper angled side neck. Heavy wall provides mechanical shock resistance.