Learn More
Ace Glass, Inc. Flask, round, 50mL, 14/20 center, 14/20 side
SDP
Catalog No. 50976849
Encompass_Preferred
50mL standard wall round bottom flask with (1) 14/20 standard taper center neck and (1) 14/20 standard taper angled side neck. Standard wall allows for more rapid heating.
Catalog No. 50-976-849 Supplier Ace Glass, Inc. Supplier No. 946408
50mL standard wall round bottom flask with (1) 14/20 standard taper center neck and (1) 14/20 standard taper angled side neck. Standard wall allows for more rapid heating.