Learn More
Ace Glass, Inc. Flask, round bottom, two neck, 5L, 24/40 center, 24/40 side
SDP
Catalog No. 50973577
Encompass_Preferred
5L two neck, round bottom flask with standard joints. Center neck and angled side neck both have 24/40 joints.
Catalog No. 50-973-577 Supplier Ace Glass, Inc. Supplier No. 692762
5L two neck, round bottom flask with standard joints. Center neck and angled side neck both have 24/40 joints.