Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 5L, 29/42 center, 24/40 sides
SDP
Catalog No. 50973809
Encompass_Preferred
5L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 29/42 joint, and angled side necks have 24/40 joints.
Catalog No. 50-973-809 Supplier Ace Glass, Inc. Supplier No. 695252
5L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 29/42 joint, and angled side necks have 24/40 joints.