Learn More
Ace Glass, Inc. Flask, round bottom, two neck, 6L, 45/50 center, 24/40 side
SDP
Catalog No. 50973579
Encompass_Preferred
6L two neck, round bottom flask with standard joints. Center neck has a 45/50 joint, and the angled side neck has a 24/40 joint.
Catalog No. 50-973-579 Supplier Ace Glass, Inc. Supplier No. 692768
6L two neck, round bottom flask with standard joints. Center neck has a 45/50 joint, and the angled side neck has a 24/40 joint.