Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 6L, 45/50 center, 29/42 sides
SDP
Catalog No. 50973673
Encompass_Preferred
6L round bottom flask with three vertical necks. Center neck has 45/50 outer joint, and side necks have 29/42 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-973-673 Supplier Ace Glass, Inc. Supplier No. 6944119
6L round bottom flask with three vertical necks. Center neck has 45/50 outer joint, and side necks have 29/42 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.