Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 6L, 45/50 center, (2) 29/42, (1) 10/30 sides
SDP
Catalog No. 50973860
Encompass_Preferred
6L round bottom flask with four necks, arranged three in-line, and one 10/30 joint at 90. Center neck has a 45/50 joint, and vertical side necks have 29/42 joints.
Catalog No. 50-973-860 Supplier Ace Glass, Inc. Supplier No. 695366
6L round bottom flask with four necks, arranged three in-line, and one 10/30 joint at 90. Center neck has a 45/50 joint, and vertical side necks have 29/42 joints.