Learn More
Ace Glass, Inc. Flask, round bottom, two neck, 6L, 45/50 center, 29/42 side
SDP
Supplier:  Ace Glass, Inc. 692770
Encompass_Preferred
6L two neck, round bottom flask with standard joints. Center neck has a 45/50 joint, and the angled side neck has a 29/42 joint.
Catalog No. 50-973-580