Learn More
Ace Glass, Inc. Flask, round bottom, three neck, 6L, 45/50 center, 45/50 sides
SDP
Catalog No. 50978964
Encompass_Preferred
6L round bottom flask with three vertical necks. Center neck and side necks have 45/50 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.
Catalog No. 50-978-964 Supplier Ace Glass, Inc. Supplier No. 6944120
6L round bottom flask with three vertical necks. Center neck and side necks have 45/50 outer joints. Flask features a flare-out directly below the center joint to facilitate cleaning and removal of stir blades.