Learn More
Ace Glass, Inc. Flask, round bottom, four neck, 6L, 55/50 center, 24/40 sides
SDP
Catalog No. 50973830
Encompass_Preferred
6L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 55/50 joint, and angled side necks have 24/40 joints.
Catalog No. 50-973-830 Supplier Ace Glass, Inc. Supplier No. 6952117
6L round bottom flask with four necks, arranged three in-line and one at 90. Center neck has a 55/50 joint, and angled side necks have 24/40 joints.