Learn More
Chemglass Life Sciences Chemglass 1000 mL Heavy-Wall 3-Neck Round Bottom Flask with Drain Valve (24/40 joints)
SDP
Catalog No. NC2319638
Encompass_Preferred
Chemglass CG-1523-V-02 is a 1,000 mL heavy-wall, three-neck round-bottom flask with an integrated drain valve. It is suited for laboratory synthesis, reflux, and applications requiring uniform heating, multi-access connections, and controlled drainage.
  • 1000 mL capacity.
  • Three necks with 24/40 outer joints.
  • Heavy-wall borosilicate glass construction for increased durability.
  • Lower outlet with extended-tip drain valve for complete drainage.
  • Sidearm uses a 1/2 in tubing fitting.
  • Round-bottom shape for uniform heating and mixing.
Catalog No. NC2319638 Supplier Chemglass Life Sciences Supplier No. CG1523V02
Chemglass CG-1523-V-02 is a 1,000 mL heavy-wall, three-neck round-bottom flask with an integrated drain valve. It is suited for laboratory synthesis, reflux, and applications requiring uniform heating, multi-access connections, and controlled drainage.
  • 1000 mL capacity.
  • Three necks with 24/40 outer joints.
  • Heavy-wall borosilicate glass construction for increased durability.
  • Lower outlet with extended-tip drain valve for complete drainage.
  • Sidearm uses a 1/2 in tubing fitting.
  • Round-bottom shape for uniform heating and mixing.