Learn More
Chemglass Life Sciences Flask, Round Bottom, 1000mL, Heavy Wall, 3-Neck, 45/50 CN - 24/40 SN's Angled 20°
SDP
Catalog No. 50873131
Encompass_Preferred
Flask, Round Bottom, 1000mL, Heavy Wall, 45/50 - 24/40, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 45/50Side neck: 24/40
Catalog No. 50-873-131 Supplier Chemglass Life Sciences Supplier No. CG152448
Flask, Round Bottom, 1000mL, Heavy Wall, 45/50 - 24/40, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 45/50Side neck: 24/40