Learn More
Chemglass Life Sciences 100mL Single Neck Round Bottom Flask (RBF) , 19/38 Outer Joint
SDP
Catalog No. 501531122
Encompass_Preferred
100mL Single Neck Round Bottom Flask, 19/38 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. 50-153-1122 Supplier Chemglass Life Sciences Supplier No. CG150604
100mL Single Neck Round Bottom Flask, 19/38 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.