Learn More
Chemglass Life Sciences 125mL Single Neck Round Bottom Flask (RBF) , 29/42 Outer Joint
SDP
Catalog No. 50873082
Encompass_Preferred
125mL Single Neck Round Bottomed Flask, 29/42 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. 50-873-082 Supplier Chemglass Life Sciences Supplier No. CG150609
125mL Single Neck Round Bottomed Flask, 29/42 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.