Learn More
Chemglass Life Sciences Flask, Round Bottom, 12000mL, Heavy Wall, 5-Neck, 45/50 CN - 24/40 SN's, Vertical
SDP
Catalog No. 501531154
Encompass_Preferred
Flask, Round Bottom, 12000mL, Heavy Wall, 45/50 - 24/40, 5-Neck, Vertical. Round bottom, HEAVY WALL, five-neck flask with standard taper outer joints on the center neck and vertical side necks.
Catalog No. 50-153-1154 Supplier Chemglass Life Sciences Supplier No. CG153510
Flask, Round Bottom, 12000mL, Heavy Wall, 45/50 - 24/40, 5-Neck, Vertical. Round bottom, HEAVY WALL, five-neck flask with standard taper outer joints on the center neck and vertical side necks.