Learn More
Ace Glass, Inc. Flask, round bottom, single neck, 1L, 35/20
SDP
Catalog No. 50973552
Encompass_Preferred
1L spherical receiving flask made of standard wall glass. Flask has a short neck and a 35/20 outer joint.
Catalog No. 50-973-552 Supplier Ace Glass, Inc. Supplier No. 690227
1L spherical receiving flask made of standard wall glass. Flask has a short neck and a 35/20 outer joint.