Learn More
Chemglass Life Sciences Round-bottom Flask, 2,000 mL, 3-Neck Heavy-Wall with Drain Valve, 24/40 Joints
SDP
Catalog No. NC2680694
Encompass_Preferred
Heavy-wall three-neck round-bottom flask with 2,000 mL capacity and an integrated bottom drain valve for controlled emptying and full agitation of the contents. Designed for laboratory use with standard taper joints for accessories and condensers.
  • Capacity: 2,000 mL.
  • Three necks with 24/40 standard taper joints.
  • Integrated bottom drain valve with extended tip for complete drainage.
  • Lower outlet sidearm with 1/2 in connection for fittings.
  • Heavy-wall glass construction for durability.
  • Clear finish for visual monitoring of contents.
Catalog No. NC2680694 Supplier Chemglass Life Sciences Supplier No. CG1523V03
Heavy-wall three-neck round-bottom flask with 2,000 mL capacity and an integrated bottom drain valve for controlled emptying and full agitation of the contents. Designed for laboratory use with standard taper joints for accessories and condensers.
  • Capacity: 2,000 mL.
  • Three necks with 24/40 standard taper joints.
  • Integrated bottom drain valve with extended tip for complete drainage.
  • Lower outlet sidearm with 1/2 in connection for fittings.
  • Heavy-wall glass construction for durability.
  • Clear finish for visual monitoring of contents.