Learn More
Chemglass Life Sciences Flask, Round Bottom, 200mL, Heavy Wall, 3-Neck, 24/40 CN - 24/40 SN's Angled 20°
SDP
Catalog No. 501531145
Encompass_Preferred
Flask, Round Bottom, 200mL, Heavy Wall, 24/40 - 24/40, 3-Neck, Angled 20. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20 angled side necks.
Catalog No. 50-153-1145 Supplier Chemglass Life Sciences Supplier No. CG152402
Flask, Round Bottom, 200mL, Heavy Wall, 24/40 - 24/40, 3-Neck, Angled 20. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20 angled side necks.