Learn More
Chemglass Life Sciences Flask, Round Bottom, 20000mL, Heavy Wall, 5-Neck, 55/50 CN - 45/50 SN's, Vertical
SDP
Catalog No. 501531155
Encompass_Preferred
Flask, Round Bottom, 22000mL, Heavy Wall, 55/50 - 45/50, 5-Neck, Vertical. Round bottom, HEAVY WALL, five-neck flask with standard taper outer joints on the center neck and vertical side necks.
Catalog No. 50-153-1155 Supplier Chemglass Life Sciences Supplier No. CG153516
Flask, Round Bottom, 22000mL, Heavy Wall, 55/50 - 45/50, 5-Neck, Vertical. Round bottom, HEAVY WALL, five-neck flask with standard taper outer joints on the center neck and vertical side necks.