Learn More
Chemglass Life Sciences 250mL Single Neck Round Bottom Flask (RBF) , 14/20 Outer Joint, #7 Chem-Thread Inlet
SDP
Catalog No. 501531139
Encompass_Preferred
250mL Single Neck Round Bottom Flask, 14/20 Outer Joint, #7 Chem-Thread Inlet. Round bottom, HEAVY WALL flask with a standard taper outer joint and a #7 Chem-Thread as a side neck for insertion of thermometers or bleed tubes having an O.D. between 4 and 7mm.
Catalog No. 50-153-1139 Supplier Chemglass Life Sciences Supplier No. CG151424
250mL Single Neck Round Bottom Flask, 14/20 Outer Joint, #7 Chem-Thread Inlet. Round bottom, HEAVY WALL flask with a standard taper outer joint and a #7 Chem-Thread as a side neck for insertion of thermometers or bleed tubes having an O.D. between 4 and 7mm.