Learn More
Chemglass Life Sciences 25mL 3-Neck Round Bottom Flask (RBF) , Minum-Ware
SDP
Catalog No. 501532690
Encompass_Preferred
25mL 3-Neck Round Bottomed Flask, Minum-Ware. Micro round bottom 3-neck flask with all 14/10 threaded outer joints. Supplied complete with open-top screw caps (MW-102-33) and Viton O-rings (MW-100-08).
Catalog No. 50-153-2690 Supplier Chemglass Life Sciences Supplier No. MW5901
25mL 3-Neck Round Bottomed Flask, Minum-Ware. Micro round bottom 3-neck flask with all 14/10 threaded outer joints. Supplied complete with open-top screw caps (MW-102-33) and Viton O-rings (MW-100-08).