Learn More
Ace Glass, Inc. Flask, round bottom, single neck, 2L, 35/25
SDP
Catalog No. 50973557
Encompass_Preferred
2L spherical receiving flask made of standard wall glass. Flask has a short neck and a 35/25 outer joint.
Catalog No. 50-973-557 Supplier Ace Glass, Inc. Supplier No. 690242
2L spherical receiving flask made of standard wall glass. Flask has a short neck and a 35/25 outer joint.