Learn More
Chemglass Life Sciences Chemglass Round Bottom Flask, 3-Neck, Heavy Wall, 3,000 mL (24/40 joints) - CG-1523-V-04
SDP
Catalog No. 502733585
Encompass_Preferred
Chemglass CG-1523-V-04 is a heavy-wall, three-neck round bottom flask with a 3,000 mL capacity and an integrated bottom drain valve. It is designed for laboratory reactions that require multiple access points, reliable drainage, and compatibility with standard taper glassware.
  • Heavy-wall borosilicate glass construction for durability and thermal resistance.
  • Three-neck configuration provides multiple access points for condensers, thermometers, and addition funnels.
  • Standard 24/40 taper joints on center and side necks for broad accessory compatibility.
  • Integrated extended-tip drain valve permits complete drainage and agitation of contents.
  • Drain valve sidearm accepts 1/2 in Swagelok fittings for secure connections.
  • 3,000 mL capacity suitable for medium-scale reactions and synthesis.
Catalog No. 50-273-3585 Supplier Chemglass Life Sciences Supplier No. CG1523V04
Chemglass CG-1523-V-04 is a heavy-wall, three-neck round bottom flask with a 3,000 mL capacity and an integrated bottom drain valve. It is designed for laboratory reactions that require multiple access points, reliable drainage, and compatibility with standard taper glassware.
  • Heavy-wall borosilicate glass construction for durability and thermal resistance.
  • Three-neck configuration provides multiple access points for condensers, thermometers, and addition funnels.
  • Standard 24/40 taper joints on center and side necks for broad accessory compatibility.
  • Integrated extended-tip drain valve permits complete drainage and agitation of contents.
  • Drain valve sidearm accepts 1/2 in Swagelok fittings for secure connections.
  • 3,000 mL capacity suitable for medium-scale reactions and synthesis.