Learn More
Chemglass Life Sciences 35mL Single Neck Round Bottom Flask (RBF) , 19/22 Outer Joint
SDP
Catalog No. 50873088
Encompass_Preferred
35mL Single Neck Round Bottomed Flask, 19/22 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. 50-873-088 Supplier Chemglass Life Sciences Supplier No. CG150632
35mL Single Neck Round Bottomed Flask, 19/22 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.