Learn More
Chemglass Life Sciences Flask, Round Bottom, 3000mL, Heavy Wall, 5-Neck, 24/40 CN - 24/40 SN's, Vertical
SDP
Catalog No. 501531153
Encompass_Preferred
Flask, Round Bottom, 3000mL, Heavy Wall, 24/40 - 24/40, 5-Neck, Vertical. Round bottom, HEAVY WALL, five-neck flask with standard taper outer joints on the center neck and vertical side necks.
Catalog No. 50-153-1153 Supplier Chemglass Life Sciences Supplier No. CG153504
Flask, Round Bottom, 3000mL, Heavy Wall, 24/40 - 24/40, 5-Neck, Vertical. Round bottom, HEAVY WALL, five-neck flask with standard taper outer joints on the center neck and vertical side necks.