Learn More
Chemglass Life Sciences Flask, Round Bottom, 5000mL, Heavy Wall, 3-Neck, 29/42 CN - 24/40 SN's Angled 20°
SDP
Catalog No. 50873130
Encompass_Preferred
Flask, Round Bottom, 5000mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/42Side neck: 24/40
Catalog No. 50-873-130 Supplier Chemglass Life Sciences Supplier No. CG152420
Flask, Round Bottom, 5000mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/42Side neck: 24/40