Learn More
Chemglass Life Sciences Flask, Round Bottom, 500mL, Heavy Wall, 4-Neck, 24/40 CN - 24/40 SN's Angled 20°
SDP
Catalog No. 50873146
Encompass_Preferred
Flask, Round Bottom, 500mL, Heavy Wall, 24/40 - 24/40, 4-Neck, Angled 20¿. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and 20¿ angled side necks.
Catalog No. 50-873-146 Supplier Chemglass Life Sciences Supplier No. CG153203
Flask, Round Bottom, 500mL, Heavy Wall, 24/40 - 24/40, 4-Neck, Angled 20¿. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and 20¿ angled side necks.