Learn More
Chemglass Life Sciences 500mL 3-Neck Round Bottom Flask (RBF) , 1-CN 45/50 Outer, 2-SN 24/40 Outer
SDP
Catalog No. 501531142
Encompass_Preferred
500mL 3-Neck Round Bottomed Flask, 1-CN 45/50 Outer, 2-SN 24/40 Outer. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and vertical side necks.
Catalog No. 50-153-1142 Supplier Chemglass Life Sciences Supplier No. CG152283
500mL 3-Neck Round Bottomed Flask, 1-CN 45/50 Outer, 2-SN 24/40 Outer. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and vertical side necks.