Learn More
Chemglass Life Sciences Flask, Round Bottom, 500mL, Heavy Wall, 3-Neck, 19/22 CN - 19/22 SN's Angled 20°
SDP
Catalog No. 501531146
Encompass_Preferred
Flask, Round Bottom, 500mL, Heavy Wall, 19/22 - 19/22, 3-Neck, Angled 20. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20 angled side necks.
Catalog No. 50-153-1146 Supplier Chemglass Life Sciences Supplier No. CG1524A16
Flask, Round Bottom, 500mL, Heavy Wall, 19/22 - 19/22, 3-Neck, Angled 20. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20 angled side necks.