Learn More
Chemglass Life Sciences 500mL Single Neck Round Bottom Flask (RBF) , 28/15 Socket Joint
SDP
Catalog No. 501531129
Encompass_Preferred
500mL Single Neck Round Bottom Flask, 28/15 Socket Joint. Round bottom, HEAVY WALL single short neck flask with a spherical socket joint.
Catalog No. 50-153-1129 Supplier Chemglass Life Sciences Supplier No. CG150807
500mL Single Neck Round Bottom Flask, 28/15 Socket Joint. Round bottom, HEAVY WALL single short neck flask with a spherical socket joint.