Learn More
Chemglass Life Sciences 50mL 3-Neck Round Bottom Flask (RBF) , Minum-Ware
SDP
Catalog No. 50574131
Encompass_Preferred
50mL 3-Neck RBF, Minum-Ware. Micro round bottom 3-neck flask with all 14/10 threaded outer joints. Supplied complete with open-top screw caps (MW-102-33) and Viton O-rings (MW-100-08).
Catalog No. 50-574-131 Supplier Chemglass Life Sciences Supplier No. MW5902
50mL 3-Neck RBF, Minum-Ware. Micro round bottom 3-neck flask with all 14/10 threaded outer joints. Supplied complete with open-top screw caps (MW-102-33) and Viton O-rings (MW-100-08).