Learn More
Ace Glass, Inc. Flask, round bottom, single neck, 50mL, 35/25
SDP
Catalog No. 50973554
Encompass_Preferred
50mL spherical receiving flask made of standard wall glass. Flask has a short neck and a 35/25 outer joint.
Catalog No. 50-973-554 Supplier Ace Glass, Inc. Supplier No. 690234
50mL spherical receiving flask made of standard wall glass. Flask has a short neck and a 35/25 outer joint.