Learn More
Chemglass Life Sciences Flask, Round Bottom, 50mL, Heavy Wall, 4-Neck, 14/20 CN - 14/20 SN's Angled 20°
SDP
Catalog No. 50873143
Encompass_Preferred
Flask, Round Bottom, 50mL, Heavy Wall, 14/20 - 14/20, 4-Neck, Angled 20¿. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and 20¿ angled side necks.
Catalog No. 50-873-143 Supplier Chemglass Life Sciences Supplier No. CG153103
Flask, Round Bottom, 50mL, Heavy Wall, 14/20 - 14/20, 4-Neck, Angled 20¿. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and 20¿ angled side necks.