Learn More
Chemglass Life Sciences 5mL Single Neck Round Bottom Flask (RBF) , 14/10 Threaded Outer Joint
SDP
Catalog No. 50873228
Encompass_Preferred
5mL Single Neck RBF, 14/10 Threaded Outer Joint. Micro round bottom flask with a 14/10 threaded outer joint. Supplied complete with an open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).
Catalog No. 50-873-228 Supplier Chemglass Life Sciences Supplier No. MW5801
5mL Single Neck RBF, 14/10 Threaded Outer Joint. Micro round bottom flask with a 14/10 threaded outer joint. Supplied complete with an open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).