Learn More
Chemglass Life Sciences 65mL Single Neck Round Bottom Flask (RBF) , 19/22 Outer Joint
SDP
Catalog No. 50873087
Encompass_Preferred
65mL Single Neck Round Bottomed Flask, 19/22 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. 50-873-087 Supplier Chemglass Life Sciences Supplier No. CG150622
65mL Single Neck Round Bottomed Flask, 19/22 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.