Learn More
Chemglass Life Sciences 6mL Single Neck Round Bottom Flask (RBF) , 19/22 Outer Joint
SDP
Catalog No. 501531124
Encompass_Preferred
6mL Single Neck Round Bottom Flask, 19/22 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. 50-153-1124 Supplier Chemglass Life Sciences Supplier No. CG150613
6mL Single Neck Round Bottom Flask, 19/22 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.