Learn More
Chemglass Life Sciences 72000mL 3-Neck Round Bottom Flask (RBF) , 1-CN 71/60 Outer, 2-SN 45/50 Outer
SDP
Catalog No. 501531141
Encompass_Preferred
72000mL 3-Neck Round Bottom Flask, 1-CN 71/60 Outer, 2-SN 45/50 Outer. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and vertical side necks.
Catalog No. 50-153-1141 Supplier Chemglass Life Sciences Supplier No. CG152280
Add to Cart
Add to Cart
Request a Quote
72000mL 3-Neck Round Bottom Flask, 1-CN 71/60 Outer, 2-SN 45/50 Outer. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and vertical side necks.