Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, single neck, 1L, 35/20
SDP
Catalog No. 50973560
Encompass_Preferred
1L heavy wall spherical receiving flask. Flask has a short neck and a 35/20 outer joint.
Catalog No. 50-973-560 Supplier Ace Glass, Inc. Supplier No. 6902227
1L heavy wall spherical receiving flask. Flask has a short neck and a 35/20 outer joint.