Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, single neck, 5L, 35/25
SDP
Catalog No. 50973567
Encompass_Preferred
5L heavy wall spherical receiving flask. Flask has a short neck and a 35/25 outer joint.
Catalog No. 50-973-567 Supplier Ace Glass, Inc. Supplier No. 6902244
5L heavy wall spherical receiving flask. Flask has a short neck and a 35/25 outer joint.