Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, 100mL, 24/40 center, #7Ace-Thred side port
SDP
Catalog No. 50973623
Encompass_Preferred
100mL heavy wall, round bottom flask with 24/40 center and #7 Ace-Thred side port. Supplied with nylon bushing and FETFE o-ring. Bushing allows for use with thermometers or bleed tubes up to 7mm O.D.
Catalog No. 50-973-623 Supplier Ace Glass, Inc. Supplier No. 6936230
100mL heavy wall, round bottom flask with 24/40 center and #7 Ace-Thred side port. Supplied with nylon bushing and FETFE o-ring. Bushing allows for use with thermometers or bleed tubes up to 7mm O.D.