Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, single neck, 125mL, 24/40
SDP
Catalog No. 50973497
Encompass_Preferred
Flask, round bottom, heavy wall, single neck, 125mL, 24/40
Catalog No. 50-973-497 Supplier Ace Glass, Inc. Supplier No. 6887232
Flask, round bottom, heavy wall, single neck, 125mL, 24/40