Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, single neck, 125mL, 29/42
SDP
Catalog No. 50973500
Encompass_Preferred
Flask, round bottom, heavy wall, single neck, 125mL, 29/42
Catalog No. 50-973-500 Supplier Ace Glass, Inc. Supplier No. 6887236
Flask, round bottom, heavy wall, single neck, 125mL, 29/42